Betacarotene for system suitability
Synonym(s):
β-Carotene, β,β-Carotene, Provitamin A
Empirical Formula (Hill Notation):
C40H56
CAS Number:
Molecular Weight:
536.87
Products
Formula Weight
Manufacturer
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing