Skip to Content
IN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Safety Data Sheets (SDS)
Certificates (COA/COO)
Quality & Regulatory
Calculators & Apps
Webinars
Back
AxySaver tube caps
Products
Product Category
caps (16)
Manufacturer
Diameter (mm)
Color
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
AXYAS13MMY
AxySaver tube caps,
diam. 13 mm, yellow
Expand
View Pricing
AXYAS16MMBK
AxySaver tube caps,
diam. 16 mm, black
Expand
View Pricing
AXYAS16MMGR
AxySaver tube caps,
diam. 16 mm, gray
Expand
View Pricing
AXYAS16MMG
AxySaver tube caps,
diam. 16 mm, green
Expand
View Pricing
AXYAS16MML
AxySaver tube caps,
diam. 16 mm, lavender
Expand
View Pricing
AXYAS16MMR
AxySaver tube caps,
diam. 16 mm, red
Expand
View Pricing
AXYAS16MMY
AxySaver tube caps,
diam. 16 mm, yellow
Expand
View Pricing
AXYAS16MMW
AxySaver tube caps,
diam. 16 mm, white
Expand
View Pricing
AXYAS16MMB
AxySaver tube caps,
diam. 16 mm, blue
Expand
View Pricing
AXYAS13MMBK
AxySaver tube caps,
diam. 13 mm, black
Expand
View Pricing
AXYAS13MMB
AxySaver tube caps,
diam. 13 mm, blue
Expand
View Pricing
AXYAS13MMGR
AxySaver tube caps,
diam. 13 mm, gray
Expand
View Pricing
AXYAS13MMG
AxySaver tube caps,
diam. 13 mm, green
Expand
View Pricing
AXYAS13MML
AxySaver tube caps,
diam. 13 mm, lavender
Expand
View Pricing
AXYAS13MMR
AxySaver tube caps,
diam. 13 mm, red
Expand
View Pricing
AXYAS13MMW
AxySaver tube caps,
diam. 13 mm, white
Expand
View Pricing