Skip to Content
IN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Safety Data Sheets (SDS)
Certificates (COA/COO)
Quality & Regulatory
Calculators & Apps
Webinars
Back
AxyCap 10 universal fit cap, 16 mm diam.
Products
Product Category
caps (8)
Manufacturer
Color
Special Grade
Natural (1)
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
AXYACPEBK
AxyCap 10 universal fit cap, 16 mm diam.,
black polyethylene
Expand
View Pricing
AXYACPEB
AxyCap 10 universal fit cap, 16 mm diam.,
blue polyethylene
Expand
View Pricing
AXYACPEGR
AxyCap 10 universal fit cap, 16 mm diam.,
gray polyethylene
Expand
View Pricing
AXYACPEG
AxyCap 10 universal fit cap, 16 mm diam.,
green polyethylene
Expand
View Pricing
AXYACPEL
AxyCap 10 universal fit cap, 16 mm diam.,
lavender polyethylene
Expand
View Pricing
AXYACPEC
AxyCap 10 universal fit cap, 16 mm diam.,
natural polyethylene
Expand
View Pricing
AXYACPER
AxyCap 10 universal fit cap, 16 mm diam.,
red polyethylene
Expand
View Pricing
AXYACPEY
AxyCap 10 universal fit cap, 16 mm diam.,
yellow polyethylene
Expand
View Pricing