Skip to Content
Merck
Anti-Tau (3-repeat isoform RD3) Antibody, clone 8E6/C11 culture supernatant, clone 8E6/C11, Upstate®

ANTI-TAU (MAPT) Antibody

Anti-Tau (3-repeat isoform RD3) Antibody, clone 8E6/C11 culture supernatant, clone 8E6/C11, Upstate®
Synonym(s):
MAPT, MTBT1, Microtubule-associated protein tau, Neurofibrillary tangle protein, Paired helical filament-tau, MAPTL, TAU, Anti-DDPAC antibody produced in rabbit, Anti-FLJ31424 antibody produced in rabbit, Anti-FTDP-17 antibody produced in rabbit, Anti-MAPTL antibody produced in rabbit, Anti-microtubule-associated protein tau antibody produced in rabbit

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-20 of 39
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2