
Anti-SP7, (N-terminal) antibody produced in rabbit

Synonym(s):
Anti-MGC126598, Anti-OSX, Anti-osterix
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing