Skip to Content
Merck
Monoclonal Anti-Actin, α-Smooth Muscle clone 1A4, ascites fluid

Anti-α-Smooth Muscle Actin (ACTA2) Antibody,

Monoclonal Anti-Actin, α-Smooth Muscle clone 1A4, ascites fluid
Synonym(s):
SMA, Monoclonal Anti-Actin, α-Smooth Muscle, Anti-ACTA2, Anti-OTTHUMP00000020042

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Application
Post Translational Modification
Species Reactivity
Technique