![Anti-SETD3 antibody produced in rabbit Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/293/066/6db03e1a-c786-4ed4-9039-4b7a7a1ffcd4/640/6db03e1a-c786-4ed4-9039-4b7a7a1ffcd4.jpg)
Anti-SETD3 antibody produced in rabbit
![Anti-SETD3 antibody produced in rabbit Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/293/066/6db03e1a-c786-4ed4-9039-4b7a7a1ffcd4/640/6db03e1a-c786-4ed4-9039-4b7a7a1ffcd4.jpg)
Synonym(s):
Anti-SET domain-containing protein 3 antibody produced in rabbit, Anti-C14orf154, Anti-DKFZp761E1415, Anti-FLJ23027, Anti-MGC87236, Anti-SET domain containing 3
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing