
Anti-SARS-CoV Spike antibody produced in rabbit

Products
Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing