![Anti-SARS-CoV-2 (COVID-19) 3CL-PRO (NSP5) antibody produced in rabbit affinity isolated antibody](/deepweb/assets/sigmaaldrich/product/images/251/960/20c3c807-6bbc-4735-a56e-aed27fb09f2a/640/20c3c807-6bbc-4735-a56e-aed27fb09f2a.jpg)
Anti-SARS-CoV-2 (COVID-19) 3CL-PRO (NSP5) antibody produced in rabbit
![Anti-SARS-CoV-2 (COVID-19) 3CL-PRO (NSP5) antibody produced in rabbit affinity isolated antibody](/deepweb/assets/sigmaaldrich/product/images/251/960/20c3c807-6bbc-4735-a56e-aed27fb09f2a/640/20c3c807-6bbc-4735-a56e-aed27fb09f2a.jpg)
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Technique
Showing 1-2 of 2