Anti-PLCG1 antibody produced in rabbit
Synonym(s):
Anti-NCKAP3, Anti-PLC-II, Anti-PLC1, Anti-PLC148, Anti-PLCgamma1, PLC-γ-1, PLC-148, PLC-II, PLCG1, Phospholipase C-II
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing