![Anti-Cytochrome c antibody produced in sheep whole antiserum](/deepweb/assets/sigmaaldrich/product/images/214/457/279d8cb8-61dd-4dac-b27c-829eefc44790/640/279d8cb8-61dd-4dac-b27c-829eefc44790.jpg)
Anti-Cytochrome c antibody produced in sheep
![Anti-Cytochrome c antibody produced in sheep whole antiserum](/deepweb/assets/sigmaaldrich/product/images/214/457/279d8cb8-61dd-4dac-b27c-829eefc44790/640/279d8cb8-61dd-4dac-b27c-829eefc44790.jpg)
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing