Anti-CSH2 antibody produced in mouse
Synonym(s):
Anti-CS2, Anti-CSB, Anti-hCSB, Anti-CS-2, Anti-hCS-B
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2