Anti-Catalase antibody produced in rabbit
Synonym(s):
CAT, CATA
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing