![Anti-B4GALNT1 antibody produced in rabbit Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/176/581/ba53b1be-01a7-440e-a2c3-6d56706f6087/640/ba53b1be-01a7-440e-a2c3-6d56706f6087.jpg)
Anti-B4GALNT1 antibody produced in rabbit
![Anti-B4GALNT1 antibody produced in rabbit Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/176/581/ba53b1be-01a7-440e-a2c3-6d56706f6087/640/ba53b1be-01a7-440e-a2c3-6d56706f6087.jpg)
Synonym(s):
Anti-β-1,4-N-Acetyl-galactosaminyl transferase 1, Anti-GALGT, Anti-GALNACT
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing