
Anthraquinone-2-sulfonic acid sodium salt monohydrate

Synonym(s):
9,10-Dihydro-9,10-dioxo-2-anthracenesulfonic acid sodium salt monohydrate, Sodium anthraquinone-2-sulfonate
Empirical Formula (Hill Notation):
C14H7NaO5S · H2O
Products
Product Category
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing