
Angiotensin II human

Synonym(s):
Angiotensin II, Human
Empirical Formula (Hill Notation):
C50H71N13O12
CAS Number:
Molecular Weight:
1046.18
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Sterility
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing