Skip to Content
Merck
AMPSO ≥99% (titration)

AMPSO

AMPSO ≥99% (titration)
Synonym(s):
N-(1,1-Dimethyl-2-hydroxyethyl)-3-amino-2-hydroxypropanesulfonic acid
Empirical Formula (Hill Notation):
C7H17NO5S
CAS Number:
Molecular Weight:
227.28
EC Number:

Products

Shipping
Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
pK Value
Purity
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing