
Amitriptyline N-oxide

Synonym(s):
Amitriptylinoxide
Empirical Formula (Hill Notation):
C20H23NO
CAS Number:
Molecular Weight:
293.40
Products
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Technique
Showing 1-1 of 1