
AmberChrom™ 1X8 Ion Exchange Resin

Synonym(s):
AmberChrom™ 1X8 chloride form
Linear Formula:
(C10H12 · C10H10 · C8H8 · C3H9N)X
CAS Number:
Products
Shipping
Product Category
Brand
pH Value
Physical Form
Separation Technique
Quality Segment
Technique
Showing 1-3 of 3