
Alternariol

Synonym(s):
3,7,9-Trihydroxy-1-methyl-6H-dibenzo[b,d]pyran-6-one, 3,7,9-Trihydroxy-1-methyl-benzo[c]chromen-6-one
Empirical Formula (Hill Notation):
C14H10O5
CAS Number:
Molecular Weight:
258.23
Products
Product Category
Brand
Formula Weight
Format
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1