Adenine hydrochloride hydrate
Synonym(s):
6-Aminopurine hydrochloride hydrate
Empirical Formula (Hill Notation):
C5H5N5 · HCl · xH2O
CAS Number:
Molecular Weight:
171.59 (anhydrous basis)
Products
Shipping
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Product Line
Purity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing