Skip to Content
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Safety Data Sheets (SDS)
Certificates (COA/COO)
Quality & Regulatory
Calculators & Apps
Webinars
Back
ACETIC ACID 2-CYCLOHEXYL-1-METHYL-PROPYL ESTER
Linear Formula:
C
12
H
22
O
2
CAS Number:
633300-29-5
Molecular Weight:
198.308
Products
Formula Weight
Special Grade
Aldrich
CPR
(3)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
S497630
ACETIC ACID 2-CYCLOHEXYL-1-METHYL-PROPYL ESTER,
Aldrich
CPR
Expand
View Pricing
S330094
ACETIC ACID 2-CYCLOHEXYL-1-METHYL-PROPYL ESTER,
Aldrich
CPR
Expand
View Pricing
S497622
ACETIC ACID 2-CYCLOHEXYL-1-METHYL-PROPYL ESTER,
Aldrich
CPR
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All