Skip to Content
Merck
Acetamide sublimed, 99%

Acetamide

Acetamide sublimed, 99%
Synonym(s):
Amide C2, Acetamide
Linear Formula:
CH3CONH2
CAS Number:
Molecular Weight:
59.07
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Research Area
Quality Segment
Markush Class
Markush Group
Showing 1-4 of 4
Compare
Product Number
Description
Pricing