
α,β-Elemolic acid

Synonym(s):
Elemolic acid, mixture of α and β, α,β-Elemadienolic acid, α,β-Elemic acid, 3α,β-Hydroxytirucallic acid, 3-Hydroxytirucalla-8,24-dien-21-oic acid
Empirical Formula (Hill Notation):
C30H48O3
Molecular Weight:
456.70
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1