Skip to Content
Merck
5-Methoxyindole-3-carboxaldehyde ≥99%

5-Methoxyindole-3-carboxaldehyde

5-Methoxyindole-3-carboxaldehyde ≥99%
Synonym(s):
3-Formyl-5-methoxyindole, NSC 521754
Empirical Formula (Hill Notation):
C10H9NO2
CAS Number:
Molecular Weight:
175.18
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing