Skip to Content
Merck
(5E,7E)-9,10-Secocholesta-5,7,10(19)-trien-3beta-ol certified reference material, pharmaceutical secondary standard

(5E,7E)-9,10-Secocholesta-5,7,10(19)-trien-3beta-ol

(5E,7E)-9,10-Secocholesta-5,7,10(19)-trien-3beta-ol certified reference material, pharmaceutical secondary standard
Synonym(s):
5,6-trans-vitamin D3, (3b,5E,7E)-9,10-Secocholesta-5,7,10(19)-trien-3-ol, calciol, cholecalciferol, colecalciferol
Empirical Formula (Hill Notation):
C27H44O
CAS Number:
Molecular Weight:
384.64

Products

Brand
Formula Weight
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing