Skip to Content
Merck
5-Chloro-[1,2,4]triazolo[4,3-a]pyridine AldrichCPR

5-Chloro-[1,2,4]triazolo[4,3-a]pyridine

5-Chloro-[1,2,4]triazolo[4,3-a]pyridine AldrichCPR
Empirical Formula (Hill Notation):
C6H4ClN3
CAS Number:
Molecular Weight:
153.57

Products

Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing