
5α-Cholestan-3-one

Synonym(s):
3-Keto-5α-cholestane
Empirical Formula (Hill Notation):
C27H46O
CAS Number:
Molecular Weight:
386.65
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1