5,10,15,20-Tetrakis(4-hydroxyphenyl)-21H,23H-porphine
Empirical Formula (Hill Notation):
C44H30N4O4
CAS Number:
Molecular Weight:
678.73
Products
Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing