Skip to Content
Merck
4-Methylpiperidine 96%

4-Methylpiperidine

4-Methylpiperidine 96%
Synonym(s):
γ-Pipecoline, 4-Pipecoline
Empirical Formula (Hill Notation):
C6H13N
CAS Number:
Molecular Weight:
99.17
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing