Skip to Content
Merck
vibrant-m

4-Hydroxy-N,N-dimethyl-α,α-4-triphenyl-1-piperidinebutanamide

vibrant-m
Synonym(s):
4-(4-Hydroxy-4-phenylpiperidin-1-yl)-N,N-dimethyl-2,2-diphenylbutanamide
Empirical Formula (Hill Notation):
C29H34N2O2
CAS Number:
Molecular Weight:
442.59

Products

Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
API Family
Reference Material Type
Special Grade