Skip to Content
Merck
4-Hydroxy-2,5-dimethyl-3(2H)-furanone analytical standard

4-Hydroxy-2,5-dimethyl-3(2H)-furanone

4-Hydroxy-2,5-dimethyl-3(2H)-furanone analytical standard
Synonym(s):
2,5-Dimethyl-4-hydroxy-3(2H)-furanone, Furaneol, Strawberry furanone
Empirical Formula (Hill Notation):
C6H8O3
CAS Number:
Molecular Weight:
128.13
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Feature
Formula Weight
Melting Point (°C)
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique