Skip to Content
Merck
4-Fluoro-7-nitrobenzofurazan BioReagent, suitable for fluorescence, ≥98.0% (HPLC)

4-Fluoro-7-nitrobenzofurazan

4-Fluoro-7-nitrobenzofurazan BioReagent, suitable for fluorescence, ≥98.0% (HPLC)
Synonym(s):
NBD-F
Empirical Formula (Hill Notation):
C6H2FN3O3
CAS Number:
Molecular Weight:
183.10

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Product Line
Purity
Quality Segment