![3-epi-25-Hydroxyvitamin D3 (6,19,19-d3) solution 50 μg/mL in ethanol, 98 atom % D, 98% (CP)](/deepweb/assets/sigmaaldrich/product/structures/404/580/16b16901-61e1-42a5-baa6-69ec7d04710a/640/16b16901-61e1-42a5-baa6-69ec7d04710a.png)
3-epi-25-Hydroxyvitamin D3 (6,19,19-d3) solution
![3-epi-25-Hydroxyvitamin D3 (6,19,19-d3) solution 50 μg/mL in ethanol, 98 atom % D, 98% (CP)](/deepweb/assets/sigmaaldrich/product/structures/404/580/16b16901-61e1-42a5-baa6-69ec7d04710a/640/16b16901-61e1-42a5-baa6-69ec7d04710a.png)
Empirical Formula (Hill Notation):
C27D3H41O2
Molecular Weight:
403.66
EC Number:
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Isotope
Application
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing