
3-(α-Acetonylbenzyl)-4-hydroxycoumarin sodium salt

Synonym(s):
4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin
Linear Formula:
C19H15O4Na
CAS Number:
Molecular Weight:
330.31
Products
Shipping
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing