Skip to Content
Merck
2-Ethylanthraquinone ≥97%

2-Ethylanthraquinone

2-Ethylanthraquinone ≥97%
Synonym(s):
2-Ethylanthraquinone
Empirical Formula (Hill Notation):
C16H12O2
CAS Number:
Molecular Weight:
236.27

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing