Skip to Content
Merck
2-Ethyl-3-methoxypyrazine ≥99%, FG

2-Ethyl-3-methoxypyrazine

2-Ethyl-3-methoxypyrazine ≥99%, FG
Empirical Formula (Hill Notation):
C7H10N2O
CAS Number:
Molecular Weight:
138.17
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing