Skip to Content
Merck
2-Adamantylamine hydrochloride 99%

2-Adamantylamine hydrochloride

2-Adamantylamine hydrochloride 99%
Synonym(s):
2-Adamantanamine hydrochloride, 2-Aminoadamantane hydrochloride
Empirical Formula (Hill Notation):
C10H17N · HCl
CAS Number:
Molecular Weight:
187.71
EC Number:

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing