
25-Hydroxyvitamin D3 monohydrate

Synonym(s):
25-Hydroxycholecalciferol
Empirical Formula (Hill Notation):
C27H44O2 · H2O
CAS Number:
Molecular Weight:
418.65
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1