![2,3-Dichloro-5,6-dicyano-p-benzoquinone 98%](/deepweb/assets/sigmaaldrich/product/structures/254/191/9261343e-a09b-4226-805a-1f7df856ba74/640/9261343e-a09b-4226-805a-1f7df856ba74.png)
2,3-Dichloro-5,6-dicyano-p-benzoquinone
![2,3-Dichloro-5,6-dicyano-p-benzoquinone 98%](/deepweb/assets/sigmaaldrich/product/structures/254/191/9261343e-a09b-4226-805a-1f7df856ba74/640/9261343e-a09b-4226-805a-1f7df856ba74.png)
Synonym(s):
4,5-Dichloro-3,6-dioxo-1,4-cyclohexadiene-1,2-dicarbonitrile, DDQ, 2,3-Dichloro-5,6-dicyano-p-benzoquinone
Linear Formula:
2,3-(Cl)2-1-(=O)C6-4-(=O)-5,6-(CN)2
CAS Number:
Molecular Weight:
227.00
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing