Skip to Content
Merck
2-[1-[(2RS)-2-(2,4-Dichlorophenyl)-2-[[(2,4-dichlorophenyl)methyl]oxy]ethyl]-1H-imidazol-3-ium-3-yl]-2-methylpropanoate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

2-[1-[(2RS)-2-(2,4-Dichlorophenyl)-2-[[(2,4-dichlorophenyl)methyl]oxy]ethyl]-1H-imidazol-3-ium-3-yl]-2-methylpropanoate

2-[1-[(2RS)-2-(2,4-Dichlorophenyl)-2-[[(2,4-dichlorophenyl)methyl]oxy]ethyl]-1H-imidazol-3-ium-3-yl]-2-methylpropanoate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Miconazole N-(2-Methyl)propanoate, Miconazole Nitrate impurity E (PhEur)
Empirical Formula (Hill Notation):
C22H20Cl4N2O3
Molecular Weight:
502.22

Products

Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade