1H-Pyrrolo[2,3-b]pyridin-5-ol
Empirical Formula (Hill Notation):
C7H6N2O
CAS Number:
Molecular Weight:
134.14
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing