![1-(4-Bromophenyl)-1H-pyrazole AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/892/814/54393d99-686b-49b9-8eff-4b546ba89de4/640/54393d99-686b-49b9-8eff-4b546ba89de4.png)
1-(4-Bromophenyl)-1H-pyrazole
![1-(4-Bromophenyl)-1H-pyrazole AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/892/814/54393d99-686b-49b9-8eff-4b546ba89de4/640/54393d99-686b-49b9-8eff-4b546ba89de4.png)
Empirical Formula (Hill Notation):
C9H7BrN2
CAS Number:
Molecular Weight:
223.07
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing