![12-Methoxycarnosic acid ≥95% (LC/MS-ELSD)](/deepweb/assets/sigmaaldrich/product/structures/273/933/e93fc491-03de-4751-9f37-3bbadfcb2e82/640/e93fc491-03de-4751-9f37-3bbadfcb2e82.png)
12-Methoxycarnosic acid
![12-Methoxycarnosic acid ≥95% (LC/MS-ELSD)](/deepweb/assets/sigmaaldrich/product/structures/273/933/e93fc491-03de-4751-9f37-3bbadfcb2e82/640/e93fc491-03de-4751-9f37-3bbadfcb2e82.png)
Synonym(s):
Carnosic acid 12-methyl ether
Empirical Formula (Hill Notation):
C21H30O4
CAS Number:
Molecular Weight:
346.46
Products
Product Category
Brand
Formula Weight
Physical Form
Research Area
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing