1-(2-Hydroxyethyl)-1,2,3,4-tetrahydro-2,2,4,7-tetramethylquinoline
Synonym(s):
1,2,3,4-Tetrahydro-2,2,4,7-tetramethyl-1-quinolineethanol
Empirical Formula (Hill Notation):
C15H23NO
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing