Product Name
Sephadex® G-150 Superfine Fractionation Range (MW)
Product Number
g15050
Product Brand
sigma
CAS Number
12774-36-6
Molecular Weight
Test | Specification |
---|---|
APPEARANCE | WHITE BEADS |
LOSS ON DRYING | NOT MORE THAN 10% |
PARTICLE SIZE DISTRIBUTION | NOT LESS THAN 80% WITHIN 20 AND 50 MICRONS |
SEPHADEX IS A REGISTERED TRADEMARK OF AMERSHAM PHARMACIA BIOTECH LIMITED OR ITS SUBSIDIARIES | |
DOCUMENT # I2509/12/19/00/3 |