Product Name
Ethyl diacetoacetate 97%
Product Number
249629
Product Brand
aldrich
CAS Number
603-69-0
Molecular Weight
172.18
Test | Specification |
---|---|
APPEARANCE | COLORLESS TO LIGHT YELLOW LIQUID |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
GAS LIQUID | 95% (MINIMUM) |
REVISED SEPTEMBER 5, 2007 BTR |