Product Name
[Adamantaneacetyl1, O-Et-D-Tyr2, Val4, Aminobutyryl6, Arg8,9]-Vasopressin ≥95% (HPLC)
Product Number
V2381
Product Brand
SIGMA
CAS Number
112465-00-6
Molecular Weight
1239.51
Test | Specification |
---|---|
Appearance (Color) | White |
Appearance (Form) | Powder |
Solubility (Color) | Colorless |
Solubility (Turbidity) | Clear |
1 mg/mL, H2O | |
% Peptide content | ≥75 |
% Purity (HPLC) | ≥95 |