Product Name
6-Benzyloxy-2-benzoxazolinone 97%
Product Number
653462
Product Brand
ALDRICH
CAS Number
158822-84-5
Molecular Weight
241.24
Test
Specification
APPEARANCEWHITE TO TAN POWDER, CRYSTALS OR CRYSTALLINE
 POWDER AND/OR CHUNKS
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
ELEMENTAL ANALYSISCARBON 67.3% - 72.1%
 NITROGEN 5.5% - 6.1%
HIGH PRESSURE LIQUID96.5% (MINIMUM)
 APPROVED MARCH 8, 2005 BTR