Product Name
5-Amino-4,6-dichloro-2-methylpyrimidine 97%
Product Number
548235
Product Brand
ALDRICH
CAS Number
39906-04-2
Molecular Weight
178.02
Test | Specification |
---|---|
APPEARANCE | WHITE TO TAN POWDER, CRYSTALS, CRYSTALLINE |
POWDER AND/OR CHUNKS | |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE. |
TITRATION | 96.5-103.5% (WITH AGNO3 AFTER O2 COMBUSTION) |
THIN-LAYER | CONSISTENT WITH 97% PURITY |
APPROVED MAY 30, 2008 RJM |